Skip to Content
Merck
All Photos(1)

Key Documents

P5494

Supelco

Protein Standard Set

analytical standard

Synonym(s):

Protein Calibration Set

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€327.00

€327.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
€327.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€327.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

contains

0.05% sodium azide as preservative

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

Packaging

Set includes 10 mL each protein standard.
  • Protein Standard, 20 mg/ml BSA
  • Protein Standard, 40 mg/ml BSA
  • Protein Standard, 60 mg/ml BSA
  • Protein Standard, 80 mg/ml BSA
  • Protein Standard, 100 mg/ml BSA
Each Standard contains 0.05% sodium azide as preservative and 0.85% w/v NaCl.

Other Notes

Bovine serum albumin in saline

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service