Skip to Content
Merck
All Photos(3)

Key Documents

P4722

Sigma-Aldrich

Pyridoxine hydrochloride

meets USP testing specifications

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€254.00
100 G
€1,480.00

€254.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€254.00
100 G
€1,480.00

About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein:
3632435
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058008
PubChem Substance ID:
NACRES:
NA.21

€254.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

98.0-102.0% dry basis

form

powder

color

white

mp

214-215 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vitamin B group is water-soluble and exists in six forms namely the pyridoxal, pyridoxine, pyridoxamine, pyridoxamine 5′-phosphate, pyridoxine 5′-phosphate and pyridoxal 5′-phosphate.[1] The food sources of vitamin B6 include grains, nuts, fruits, vegetables and meat.[2]

Application

Pyridoxine hydrochloride has been used to test its neuroprotective functionality in traumatic brain injury (TBI) rats.[3] It has also been used as a reference standard to quantify vitamin B6 in feed and digesta samples using high-performance liquid chromatography(HPLC).[4]

Biochem/physiol Actions

Pyridoxine plays a key role in metabolism both carbohydrates and amino acids.[2] It supports brain development and its deficiency levels may lead to depletion of γ-aminobutyric acid (GABA) levels.[5] Pyridoxine catalyzes the neurotransmitter biosynthesis like dopamine and serotonin.[5] Pyridoxine in excess or deficient may lead to neuropathy.[6]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nutritional neuropathies
Hammond N, et al.
Neurologic Clinics, 31(2), 477-477 (2013)
B Vitamins in the nervous system: Current knowledge of the biochemical modes of action and synergies of thiamine, pyridoxine, and cobalamin
Calderon-Ospina CA, et al.
CNS Neuroscience & Therapeutics (2019)
Pyridoxine administration improves behavioral and anatomical outcome after unilateral contusion injury in the rat
Kuypers NJ and Hoane MR
Journal of Neurotrauma, 27(7), 1275-1282 (2010)
Effects of forage family on apparent ruminal synthesis of B vitamins in lactating dairy cows
Castagnino DS, et al.
Journal of Dairy Science, 99(3), 1884-1894 (2016)
Vitamin B6 and its role in cell metabolism and physiology
Parra M, et al.
Cells, 7(7), 84-84 (2018)

Articles

Vitamin B6's impact on serum-free cell culture systems is explored for protein manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service