Skip to Content
Merck
All Photos(1)

Key Documents

P4554

Supelco

Proparacaine hydrochloride

analytical standard

Synonym(s):

Proxymetacaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26N2O3 · HCl
CAS Number:
Molecular Weight:
330.85
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.CCCOc1ccc(cc1N)C(=O)OCCN(CC)CC

InChI

1S/C16H26N2O3.ClH/c1-4-10-20-15-8-7-13(12-14(15)17)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H

InChI key

BFUUJUGQJUTPAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proparacaine hydrochloride is a topical ocular anesthetic[1], which is commonly used in pediatric patients.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Topical ophthalmic anesthetic

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pediatric Cataract Surgery: Techniques, Complications, and Management (2005)
Antonio Micali et al.
Ophthalmology, 121(6), 1164-1173 (2014-02-05)
To demonstrate the corneal morphologic aspects obtained with in vivo confocal microscopy (CM) and light and electron microscopy of specimens obtained from the same patients with macular corneal dystrophy (MCD). Case series. Five consecutive patients affected by MCD undergoing penetrating keratoplasty
Ditipriya R Mukhopadhyay et al.
Journal of cataract and refractive surgery, 37(5), 907-913 (2011-03-23)
To assess the effect of a combination of proparacaine 0.50%-sodium fluorescein 0.25% and ultrasound (US) pachymetry on central and midperipheral corneal thickness. School of Optometry and Vision Sciences, Cardiff University, Cardiff, Wales, United Kingdom. Case series. Topographic measurements of corneal
Gregory R Blaha et al.
Retina (Philadelphia, Pa.), 31(3), 535-539 (2010-11-26)
To compare the effectiveness of four different anesthetic methods for intravitreal injection. Twenty-four patients each received four intravitreal injections using each of four types of anesthesia (proparacaine, tetracaine, lidocaine pledget, and subconjunctival injection of lidocaine) in a prospective, masked, randomized
Mani Baskaran et al.
Investigative ophthalmology & visual science, 55(11), 7669-7673 (2014-10-23)
To evaluate a novel software capable of automatically grading angle closure on EyeCam angle images in comparison with manual grading of images, with gonioscopy as the reference standard. In this hospital-based, prospective study, subjects underwent gonioscopy by a single observer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service