Skip to Content
Merck
All Photos(1)

Key Documents

P2698000

Prednicarbate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(11β)-17-[(Ethoxycarbonyl)oxy]-11-hydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36O8
CAS Number:
Molecular Weight:
488.57
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

prednicarbate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O(C1(C2(C(C3C(C4(C(=CC(=O)C=C4)CC3)C)C(C2)O)CC1)C)C(=O)COC(=O)CC)C(=O)OCC

InChI

1S/C27H36O8/c1-5-22(31)34-15-21(30)27(35-24(32)33-6-2)12-10-19-18-8-7-16-13-17(28)9-11-25(16,3)23(18)20(29)14-26(19,27)4/h9,11,13,18-20,23,29H,5-8,10,12,14-15H2,1-4H3

InChI key

FNPXMHRZILFCKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prednicarbate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Lombardi Borgia et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 68(2), 380-389 (2007-06-19)
Reconstructed human epidermis is a useful tool for in vitro skin absorption studies of chemical compounds. If this may hold true also for topical dermatics, we investigated the glucocorticoid prednicarbate applied by two sets (innovator and generic) of cream, ointment
Pterygium unguis formation in porokeratosis of Mibelli.
D S Kim et al.
The British journal of dermatology, 156(6), 1384-1385 (2007-04-27)
Sonsoles Yáñez et al.
Dermatology (Basel, Switzerland), 208(2), 167-170 (2004-04-02)
Generalized haemorrhagic lichen nitidus is rare. To our knowledge, this form of presentation has only been reported once. To describe a new case of generalized haemorrhagic lichen nitidus simulating a pigmented purpuric dermatosis. We document a 24-year-old man who presented
M Juenger et al.
Current medical research and opinion, 22(4), 739-750 (2006-05-11)
Patients with atopic dermatitis (AD) have an increased tendency to develop bacterial skin infections. Colonization with Staphylococcus aureus is known to be a major trigger and might also play a pathophysiological role. Because of their antiseptic action, silver-coated textiles suppress
Kerstin Katzer et al.
European journal of dermatology : EJD, 20(1), 82-84 (2009-10-03)
The epidermal growth factor receptor (EGFR) inhibitor cetuximab is used to treat advanced stages of solid tumors. Acneiform skin eruptions, the most common side effects of cetuximab, stigmatize the patient in daily life and may lower compliance. As oral treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service