Skip to Content
Merck
All Photos(1)

Key Documents

P1890000

Piperacillin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Piperacillin monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N5O7S · H2O
CAS Number:
Molecular Weight:
535.57
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

piperacillin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

S1[C@H]2N([C@H](C1(C)C)C(=O)O)C(=O)[C@H]2NC(=O)[C@H](NC(=O)N4CCN(C(=O)C4=O)CC)c3ccccc3.O

InChI

1S/C23H27N5O7S.H2O/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28;/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34);1H2/t13-,14-,15+,20-;/m1./s1

InChI key

NBXPLBPWMYNZTC-IDYPWDAWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piperacillin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian D VanScoy et al.
Antimicrobial agents and chemotherapy, 58(10), 6024-6031 (2014-07-30)
It is important to understand the relationship between antibiotic exposure and the selection of drug resistance in the context of therapy exposure. We sought to identify the ceftolozane-tazobactam exposure necessary to prevent the amplification of drug-resistant bacterial subpopulations in a
Ashley W Sturm et al.
Pharmacotherapy, 34(1), 28-35 (2013-07-19)
To evaluate the steady-state pharmacokinetic and pharmacodynamic parameters of piperacillin in morbidly obese, surgical intensive care patients. Open-label single-center prospective study. Level I trauma center and university-affiliated teaching institution. Nine morbidly obese (body mass index [BMI] 40.0 kg/m² or higher)
Sangil Jeon et al.
Antimicrobial agents and chemotherapy, 58(7), 3744-3751 (2014-04-23)
Piperacillin in combination with tazobactam, a β-lactamase inhibitor, is a commonly used intravenous antibiotic for the empirical treatment of infection in intensive care patients, including burn patients. The purpose of this study was to develop a population pharmacokinetic (PK) model
Raquel Cavalcanti Dantas et al.
Journal of medical microbiology, 63(Pt 12), 1679-1687 (2014-09-28)
The rates of multidrug-resistant, extensively drug-resistant and pandrug-resistant isolates amongst non-fermenting Gram-negative bacilli, particularly Pseudomonas aeruginosa, have risen worldwide. The clinical consequence of resistance and the impact of adverse treatment on the outcome of patients with P. aeruginosa bacteraemia remain
Jessica Krezdorn et al.
Journal of medical microbiology, 63(Pt 7), 945-955 (2014-06-15)
The aim of this study was to compare the inhibitory effect of antibiotic combinations in vitro with efficacy in Galleria mellonella larvae in vivo to identify efficacious combinations that target Pseudomonas aeruginosa. P. aeruginosa NCTC 13437, a multidrug-resistant strain resistant

Questions

  1. I can not find any information about the reconstitution. How much and what has to be added to the powder?

    1 answer
    1. Product reference P1890000 Piperacillin is an European Pharmacopoeia (EP) Reference Standard as indicated in the product web page. The use of primary reference standards is prescribed by the issuing pharmacopeia.
      EP Materials are supplied exclusively as European Pharmacopoeia Chemical Reference Substances, Biological Reference Preparations or Reference Spectra (Ph. Eur. CRS, BRP or RS) for use as reference standards in tests and assays carried out in accordance with the official methods of the European Pharmacopoeia (Ph. Eur.) and for no other purpose.

      Here is how to access the information for part number P1890000:
      1) Access the EDQM website by clicking on the blue link "EDQM reference substance catalog" in the "Description" section of the product web page.
      https://crs.edqm.eu/
      2) Select "Catalog code" and enter the reference of the desired standard (P1890000)
      3) Click on the reference in blue

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service