Skip to Content
Merck
All Photos(3)

Documents

P0963

Sigma-Aldrich

Potassium oxalate monohydrate

BioXtra, ≥98.5%

Synonym(s):

Ethanedioic acid, Oxalic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOK)2 · H2O
CAS Number:
Molecular Weight:
184.23
Beilstein:
3752576
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.31

product line

BioXtra

Quality Level

Assay

≥98.5%

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.01% Insoluble matter

solubility

H2O: 0.1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%

cation traces

Al: ≤0.0005%
Ca: ≤0.002%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.002%
NH4+: ≤0.002%
Na: ≤0.02%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.[K+].[K+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • N self-doped hierarchically porous carbon derived from biomass as an efficient adsorbent for the removal of tetracycline antibiotics.: This study explores the use of nitrogen self-doped hierarchically porous carbon, derived from biomass, in removing tetracycline antibiotics from aqueous solutions. Potassium oxalate monohydrate is involved in the synthesis process, contributing to the material′s enhanced adsorption capacity. (Wang et al., 2022).

  • The influence of additives and metal rods on the nucleation and growth of gas hydrates.: This study investigates how different additives, including potassium oxalate monohydrate, and metal rods influence the nucleation and growth of gas hydrates. The findings have significant implications for the development of methods to control hydrate formation in pipelines and other industrial settings. (Li et al., 2005).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sosvorová et al.
Physiological research, 63(4), 521-527 (2014-06-09)
Increased homocysteine levels in serum are typical features of neurodegenerative brain diseases including hydrocephalus. The most frequent therapeutic approach consists of the insertion of a shunt, connecting the brain ventricles to an alternative drainage site. To decide whether the patient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service