Skip to Content
Merck
All Photos(1)

Key Documents

O5380

Supelco

Oxethazaine

analytical standard

Synonym(s):

2-Hydroxyethyliminobis(N-[α,α-dimethylphenethyl]-N-methylacetamide), Oxetacaine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€148.00

€148.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€148.00

About This Item

Empirical Formula (Hill Notation):
C28H41N3O3
CAS Number:
Molecular Weight:
467.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€148.00


Estimated to ship on28 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN(C(=O)CN(CCO)CC(=O)N(C)C(C)(C)Cc1ccccc1)C(C)(C)Cc2ccccc2

InChI

1S/C28H41N3O3/c1-27(2,19-23-13-9-7-10-14-23)29(5)25(33)21-31(17-18-32)22-26(34)30(6)28(3,4)20-24-15-11-8-12-16-24/h7-16,32H,17-22H2,1-6H3

InChI key

FTLDJPRFCGDUFH-UHFFFAOYSA-N

General description

Oxethazaine is a topical anesthetic, administered as an antacid for treating pain associated with duodenal ulcer, esophagitis and gastritis.[1]

Application

Oxethazaine may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Yasuno et al.
Biological & pharmaceutical bulletin, 21(12), 1294-1299 (1999-01-09)
Oxethazaine (OXZ), a potent topical anesthetic, was found to induce red blood cell (RBC) lysis in vitro, depending upon concentrations of OXZ, RBC and Ca2+. In a 2% RBC suspension, 100 microM OXZ caused almost complete hemolysis in the presence
A Zanni et al.
Current medical research and opinion, 10(2), 128-134 (1986-01-01)
A double-blind study was carried out in 40 patients with typical ulcer pain and suffering from gastric or duodenal ulcer to compare the pain-relieving efficacy of oxethazaine/magnesium and aluminium hydroxide mixture with that of the antacid combination alone. Patients took
Yasusuke Masuda
Journal of pharmacological sciences, 100(3), 167-174 (2006-03-07)
The liver has an intricate microvascular system that allows homogenous perfusion throughout the organ. However, the regulatory mechanisms of intrahepatic circulation are still unclear, and the effects of drugs on this system have rarely been reported. Oxethazaine, a topical anesthetic
[Resorption of cyclosporin administered by oral route to renal transplant patients. Influence of breakfast and an antacid drug].
F Penouil et al.
Therapie, 43(1), 15-19 (1988-01-01)
T Takagi et al.
The American journal of physiology, 255(4 Pt 1), G522-G528 (1988-10-01)
Celiac and superior mesenteric arterial blood flows were measured simultaneously in conscious beagle dogs. The responses to food were completely different between the two arteries. Celiac flow increased quickly to 180-200% of the control 2 min after food but started

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service