Skip to Content
Merck
All Photos(1)

Documents

O1002

Sigma-Aldrich

Oxacillin sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H18N3O5SNa · H2O
CAS Number:
Molecular Weight:
441.43
Beilstein:
4287093
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

solubility

water: 50-52 mg/mL, clear to very slightly hazy, colorless to very faintly yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].[H]O[H].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)c3c(C)onc3-c4ccccc4)C([O-])=O

InChI

1S/C19H19N3O5S.Na.H2O/c1-9-11(12(21-27-9)10-7-5-4-6-8-10)15(23)20-13-16(24)22-14(18(25)26)19(2,3)28-17(13)22;;/h4-8,13-14,17H,1-3H3,(H,20,23)(H,25,26);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1

InChI key

ZVIYWUUZWWBNMB-VICXVTCVSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oxacillin sodium salt monohydrate has been used in penicillin (PEN) standards. In turbidimetric method, oxacillin sodium salt monohydrate has been used in the medium with antibiotic.

Biochem/physiol Actions

Oxacillin and its derivatives are released through urine with the help of glomerular filtration and tubular secretion. It can be used in nursing mothers. Oxacillin is used in the treatment of methicillin-susceptible S. aureus (MSSA) infections. It shows resistance to penicillinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Washington Manual Infectious Diseases Subspecialty Consult, 352-352 (2005)
Drugs and Lactation Database (LactMed) (2006)
An easy-to-use, rapid and inexpensive method to determine methicillin resistance in Staphylococcus aureus.
Gocmen JS, et al.
Journal of Clinical and Experimental Investigations, 7(3), 225-251 (2016)
Daniel Vázquez-Sánchez et al.
International journal of food microbiology, 157(2), 286-296 (2012-06-19)
A total of 298 fishery products purchased from retail outlets in Galicia (NW Spain) between January 2008 and May 2009 were analyzed for the presence of Staphylococcus aureus. S. aureus was detected in a significant proportion of products (~25%). Incidence
Yuanli Liu et al.
Antimicrobial agents and chemotherapy, 56(11), 6048-6050 (2012-09-06)
Perturbation of hydroxyl radical accumulation by subinhibitory concentrations of 2,2'-bipyridyl plus thiourea protects Escherichia coli from being killed by 3 lethal antimicrobial classes. Here, we show that 2,2'-bipyridyl plus thiourea delays and/or reduces antimicrobial killing of Staphylococcus aureus by daptomycin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service