NIST994
Isotopic standard for gallium
NIST® SRM® 994
Synonym(s):
Gallium
About This Item
Recommended Products
grade
certified reference material
Quality Level
isotopic purity
69Ga
71Ga
form
(metallic liquid or solid (ambient temp dependent))
packaging
pkg of 0.25 g
manufacturer/tradename
NIST®
resistivity
25.795 μΩ-cm, 30°C
technique(s)
mass spectrometry (MS): suitable
bp
2403 °C (lit.)
mp
29.8 °C (lit.)
density
5.904 g/mL at 25 °C (lit.)
format
matrix material
SMILES string
[Ga]
InChI
1S/Ga
InChI key
GYHNNYVSQQEPJS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 994_cert
SRM 994_SDS
Other Notes
Gallium-69, Gallium-71
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service