Skip to Content
Merck
All Photos(2)

Documents

NIST973

Boric acid

NIST® SRM® 973, acidimetric standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

vapor pressure

2.6 mmHg ( 20 °C)

packaging

pkg of 100 g

manufacturer/tradename

NIST®

mp

160 °C (dec.) (lit.)

format

neat

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is certified as a chemical of known assay and is intended for use as a primary acidimetric standard. The SRM consists of highly purified boric acid (H3BO3) in a 100 g bottle. For more information, please refer to the SDS and COA.

SRM 973_cert

SRM 973_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Boric Acid

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service