Skip to Content
Merck
All Photos(1)

Key Documents

NIST8391

Human DNA for Whole-Genome Variant Assessment (Son of Eastern European Ashkenazi Jewish Ancestry) (HG-002)

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

0.05 G
€1,000.00
10 EA
€4,080.00

€1,000.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.05 G
€1,000.00
10 EA
€4,080.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€1,000.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 0.05 g (1 vial)
pkg of 10 × 0.05 g (10 vials)

manufacturer/tradename

NIST®

application(s)

genomic analysis

storage temp.

−20°C

General description

The Human DNA for Whole-Genome Variant Assessment Reference Material (RM) is intended for validation, optimization, and process evaluation purposes. It consists of a male human whole genome sample of Eastern European Ashkenazi Jewish ancestry from the Personal Genome Project (ID huAA53E0), and it can be used to assess the performance of variant calling from genome sequencing. A unit of RM 8391 consists of a vial containing human genomic DNA extracted from a single large growth of human lymphoblastoid cell line GM24385 (labeled as HG-002) from the Coriell Institute for Medical Research (Camden, NJ). The vial contains approximately 10 μg of genomic DNA in TE buffer (10 mM TRIS, 1 mM EDTA, pH 8.0)

SRM8391_cert

SRM8391_SDS

Application

Human DNA for Whole-Genome Variant Assessment Reference Material is intended for assessing the performance of human whole genome sequencing variant calling by obtaining estimates of true positives, false positives, and false negatives. Sequencing applications could include:

  • whole genome sequencing
  • whole exome sequencing
  • targeted sequencing such as gene panels

Features and Benefits

  • This genomic DNA Reference Material (RM) is easy to process and analyze.
  • This RM contains isolated DNA rather than live cells.
  • An extensive report of the investigation is available through NIST.
  • Information values are provided for single nucleotide variations (SNVs), small insertions and deletions (indels), and homozygous reference genotypes.

Other Notes

  • RM 8391 is stored at –20 °C but will be shipped with freezer packs and may not arrive frozen
  • Details on expiration, storage, safety, usage, and source are provided in the NIST certificate
  • Information on biomaterials, disposal, and transport is available in the SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lesley M Chapman et al.
PLoS computational biology, 16(6), e1007933-e1007933 (2020-06-20)
A high quality benchmark for small variants encompassing 88 to 90% of the reference genome has been developed for seven Genome in a Bottle (GIAB) reference samples. However a reliable benchmark for large indels and structural variants (SVs) is more
A diploid assembly-based benchmark for variants in the major histocompatibility complex
Chen-Shan Chin, et al.
Nature Communications, 11 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service