Skip to Content
Merck
All Photos(1)

Key Documents

NIST8037

Krill Oil

NIST®SRM®

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 4.5 ML
€814.00

€814.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
3 X 4.5 ML
€814.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€814.00


Estimated to ship on30 May 2025


Request a Bulk Order

form

oil

manufacturer/tradename

NIST®

storage temp.

−20°C

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service