The set of standard reference materials, NIST482, comprises six wires, each with a diameter of approximately 0.5 mm and a length of approximately 5 cm.
NIST482
Gold-copper wires for microprobe analysis
NIST® SRM® 482
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
wire
packaging
pkg of set (6)
manufacturer/tradename
NIST®
application(s)
general analytical
General description
SRM 482_cert
SRM 482_SDS
Other Notes
Gold (Au), Copper (Cu)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the diameter of the Product NIST482?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service