NIST3139A
Phosphorus (P) standard solution
NIST® SRM® 3139a
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
Related Categories
General description
SRM 3139A_cert
SRM 3139A_SDS
Application
Features and Benefits
- The certified value of the SRM was determined by calculating the mass fraction values of phosphorus, which were obtained through two methods:
- Gravimetric preparation
- Inductively coupled plasma optical emission spectrometry (ICP-OES)
- The certified value is metrologically traceable to the SI unit of mass fraction, expressed in units of milligrams per gram.
- The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.
Other Notes
Phosphorus (P)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service