Skip to Content
Merck
All Photos(2)

Key Documents

NIST2900

Ethanol-water solution

NIST® SRM® 2900, nominal 95.6% by mass

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SRM 2900 comprises five 10 milliliter ampoules, each containing approximately 10 mL of solution at an expected concentration of 95.6% by mass.



SRM 2900_cert

SRM 2900_SDS

Application

Ethanol-water solution standard reference material (SRM) is primarily intended for calibrating instruments and techniques used in determining ethanol concentrations in water-based media.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of Ethanol.
  • The certified value is obtained from the gravimetric preparation of the solution and from the analytical results determined using gas chromatography.
  • The certified value is metrologically traceable to the SI unit of mass, expressed as a percent.
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Ethanol (C2H6O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup - (in analogy to similar products)

Flash Point(C)

9.7 °C - closed cup - (in analogy to similar products)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service