Skip to Content
Merck
All Photos(1)

Documents

NIST2584

Trace elements in indoor dust

NIST® SRM® 2584

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 8 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 2584 contains 8 grams of particulate material, with more than 99% able to pass through a 100 µm (No. 145) sieve.

SRM 2584_cert

SRM 2584_SDS

Application

The SRM is suitable for use in the evaluation of methods and calibration of apparatus measuring the content of lead and other trace elements in dust.

Features and Benefits

  • Accompanied with a certificate documenting NIST-certified mass fraction values for total elements in indoor dust.
  • The reference values of elements are reported as mass fraction.
  • Certified values are metrologically traceable to the International System of Units (SI) unit of mass fraction expressed as milligrams. per kilogram on a dry-mass basis.
  • The NIST certificate is provided with period of validity, use, storage, handling, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Arsenic (As), Cadmium (Cd), Chromium (Cr), Lead (Pb), Mercury (Hg)

Reference/Informational value is provided for the following analytes.
Aluminum (Al), Calcium (Ca), Iron (Fe), Potassium (K), Lanthanum (La), Magnesium (Mg), Sodium (Na), Phosphorus (P), Titanium (Ti), Zinc (Zn), Antimony (Sb), Nickel (Ni), Barium (Ba), Niobium (Nb), Beryllium (Be), Rubidium (Rb), Bismuth (Bi), Scandium (Sc), Cerium (Ce), Selenium (Se), Cesium (Cs), Silicon (Si), Cobalt (Co), Strontium (Sr), Copper (Cu), Thorium (Th), Gallium (Ga), Uranium (U), Lithium (Li), Vanadium (V), Manganese (Mn), Yttrium (Y), Molybdenum (Mo)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service