Skip to Content
Merck
All Photos(1)

Documents

NIST2386

Avacado Powder

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 50 mg

manufacturer/tradename

NIST®

application(s)

agriculture

format

matrix material

General description

One unit of Avocado Powder, SRM 2386 contains five packets, each with around ~10 grams of freeze-dried avocado powder.
For more information, please refer to the SDS and COA.
SRM 2386_cert
SRM 2386_SDS

Application
Avocado Powder SRM is intended for evaluating methods used to determine vitamins, elements, proximates, amino acids, and fatty acids in freeze-dried avocado powder and other materials of a similar matrix.
Also used for quality assurance, including assigning values for in-house control materials.

Features and Benefits

  • Certified mass fraction values for various element are provided on a dry-mass basisand values are metrologicallytraceable to the SI unit for mass and estimated using purity evaluations and gravimetric process, expressed asmilligrams analyte per kilogram sample (mg/kg).
  • Accompaniedwith non-certified values presented as the best estimates of true values and aremetrologically traceable to results obtained from NIST measurement processusing standard methods.
  • Non-certified values are ideal for method harmonization, methoddevelopment, and process control.
  • Expirationcertification, source, instructions for storage and use are provided on the NIST certificate.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service