Skip to Content
Merck
All Photos(1)

Documents

NIST1171

Stainless Steel

NIST® SRM® 1171, Cr 17 - Ni 11 - Ti 0.3 (AISI 321)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

technique(s)

XRF: suitable
optical emission spectroscopy (OES): suitable

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 1171 consists of an annealed solid disk measuring 31 mm×19 mm.



SRM 1171_cert

SRM 1171_SDS

Application

Stainless steel standard reference material(SRM) is primarily intended for application in optical emission and X-ray fluorescence spectroscopic analytical methods.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified values of stainless-steel mass fraction and non-certified values of cobalt mass fraction.
  • Information values for elements are reported as mass fractions.
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Certified for the analytes listed below. Carbon (C), Chromium (Cr), Copper (Cu), Manganese (Mn), Molybdinum (Mo), Nickel (Ni), Silicon (Si), Titanium (Ti)

Reference/Informational value is provided for the following analytes.
Cobalt (Co), Phosphorus (P), Sulfur (S), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service