Skip to Content
Merck
All Photos(1)

Key Documents

N1391

Supelco

Nafronyl oxalate salt

analytical standard

Synonym(s):

α-(1-Naphthylmethyl)-2-tetrahydrofuranpropionic acid diethylaminoethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€88.70

€88.70


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
€88.70

About This Item

Empirical Formula (Hill Notation):
C24H33NO3 · C2H2O4
CAS Number:
Molecular Weight:
473.56
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€88.70


Estimated to ship on15 April 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(=O)C(O)=O.CCN(CC)CCOC(=O)C(CC1CCCO1)Cc2cccc3ccccc23

InChI

1S/C24H33NO3.C2H2O4/c1-3-25(4-2)14-16-28-24(26)21(18-22-12-8-15-27-22)17-20-11-7-10-19-9-5-6-13-23(19)20;3-1(4)2(5)6/h5-7,9-11,13,21-22H,3-4,8,12,14-18H2,1-2H3;(H,3,4)(H,5,6)

InChI key

SSAJNPNVUYMUCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Vasodilator

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service