Skip to Content
Merck
All Photos(1)

Key Documents

M3157

Supelco

Minaprine dihydrochloride

analytical standard

Synonym(s):

N-(4-Methyl-6-phenyl-3-pyridazinyl)-4-morpholineethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N4O · 2HCl
CAS Number:
Molecular Weight:
371.30
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Cl.Cc1cc(nnc1NCCN2CCOCC2)-c3ccccc3

InChI

1S/C17H22N4O.2ClH/c1-14-13-16(15-5-3-2-4-6-15)19-20-17(14)18-7-8-21-9-11-22-12-10-21;;/h2-6,13H,7-12H2,1H3,(H,18,20);2*1H

InChI key

GNUCGROXDZMCJI-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Antidepressant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Lorenzini et al.
Pharmacology, biochemistry, and behavior, 45(2), 481-485 (1993-06-01)
The nootropic activity of 3-(2-morpholino-ethylamino)-4-methyl-6-phenyl-pyridazine dihydrochloride (minaprine) has been investigated in intact male, adult Long Evans rats by means of an active avoidance paradigm. In the light-dark box apparatus, the rat had to learn the active avoidance response of going
A Imperato et al.
Neuroscience letters, 166(1), 69-72 (1994-01-17)
Subchronic (5 mg/kg daily for 9 consecutive days) but not acute minaprine treatment enhanced in vivo dopamine release in the limbic part of the striatum of rats as revealed by intracerebral microdialysis. Moreover, the same subchronic treatment with minaprine reduced
J G Edwards et al.
British journal of clinical pharmacology, 42(4), 491-498 (1996-10-01)
1. A double-blind, multicentre study comparing the efficacy and safety of minaprine and amitriptyline in patients with major depression was carried out. Five hundred and thirty-one patients were randomly assigned to treatment with either minaprine 100 mg, 200 mg or
T R Norman et al.
The Australian and New Zealand journal of psychiatry, 26(1), 73-81 (1992-03-01)
A review of the clinical efficacy of four structurally distinct antidepressant drugs is presented. Their antidepressant activity can be rationalised within current pharmacological hypotheses of drug action, despite markedly different effects on "in vitro" testing. Fluoxetine, a specific serotonin re-uptake
S Cabib et al.
Psychopharmacology, 121(1), 73-80 (1995-09-01)
Different types of clinically effective antidepressants prevent the behavioral effects of experimental stress, and some of these treatments affect mesolimbic dopamine (DA) functioning. Animal studies have demonstrated that repeated psychostimulant administration and repeated or chronic stressful experiences also affect mesolimbic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service