Skip to Content
Merck
All Photos(1)

Documents

M2250

Sigma-Aldrich

D-(+)-Maltose monohydrate

Type II, ≥95%

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

corn

Quality Level

type

Type II

Assay

≥95%

form

powder

optical activity

[α]20/D 129 to 131 °, c = 4% (w/v) in water + trace NH4OH

impurities

≤1.5% Glucose
≤5% Maltotriose

color

white

useful pH range

5.0-7 (25 °C, 180 g/L)

mp

125.13 °C (1013.25 hPa)

solubility

180 g/L at 20 °C

storage temp.

room temp

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Evaluation of the variations in chemical and microbiological properties of the sourdoughs produced with selected lactic acid bacteria strains during fermentation: This research explores the use of D-(+)-Maltose monohydrate in fermentative processes, highlighting its impact on the chemical and microbial dynamics in food technology, pertinent for industrial biosynthesis applications in the life sciences sector (Boyaci Gunduz et al., 2022).

Biochem/physiol Actions

Maltose is a disaccharide containing two glucose molecules with an α(1→4) glycosidic linkage. Maltose can be derived from starch in food through the action of amylase. Maltose can be found in many food products, including beer, cereals, and pasta.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wesley D Black
PloS one, 15(12), e0238901-e0238901 (2020-12-19)
Despite the recent increase in interest in indoor air quality regarding mould, there is no universally accepted standard media for the detection of airborne fungi, nor verification of many commonly used techniques. Commonly used media including malt-extract agar (MEA), Sabouraud
Vikas A Tillu et al.
Nature communications, 12(1), 931-931 (2021-02-12)
Caveolae are spherically shaped nanodomains of the plasma membrane, generated by cooperative assembly of caveolin and cavin proteins. Cavins are cytosolic peripheral membrane proteins with negatively charged intrinsically disordered regions that flank positively charged α-helical regions. Here, we show that
S K Aoki et al.
Journal of bacteriology, 191(6), 1777-1786 (2009-01-07)
Contact-dependent growth inhibition (CDI) is a mechanism identified in Escherichia coli by which bacteria expressing two-partner secretion proteins encoded by cdiA and cdiB bind to BamA in the outer membranes of target cells and inhibit their growth. A third gene
Lichao Chen et al.
Developmental cell, 53(4), 444-457 (2020-04-25)
The redox-based protein S-nitrosylation is a conserved mechanism modulating nitric oxide (NO) signaling and has been considered mainly as a non-enzymatic reaction. S-nitrosylation is regulated by the intracellular NO level that is tightly controlled by S-nitrosoglutathione reductase (GSNOR). However, the
Dengjun He et al.
Journal of separation science, 38(13), 2229-2237 (2015-04-16)
Trehalose, a nonreducing disaccharide, has been extensively applied to food, cosmetics, and pharmaceutical goods. The resultant solution of trehalose prepared by enzymatic methods includes high amounts of maltose. However, it is quite difficult to separate maltose and trehalose on an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service