Skip to Content
Merck
All Photos(1)

Documents

M0605000

Metformin hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,1-Dimethylbiguanide hydrochloride, Metformin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHC(=NH)N(CH3)2 · HCl
CAS Number:
Molecular Weight:
165.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference standard

API family

metformin

manufacturer/tradename

EDQM

mp

223-226 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CN(C)C(=N)NC(N)=N

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H

InChI key

OETHQSJEHLVLGH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metformin hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity. Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yen-Hsiang Chang et al.
Frontiers in endocrinology, 11, 445-445 (2020-08-28)
Background: Plenty of evidence suggested that chronic low-grade inflammation triggered by innate immunity activation contributes to the pathogenesis of type 2 diabetes (T2D). Using the trans-mitochondrial cybrid cell model, we have demonstrated that mitochondria independently take part in the pathological
Emi Kawakita et al.
Molecular cancer research : MCR, 19(1), 61-73 (2020-10-01)
The biological influence of antidiabetic drugs on cancer cells and diabetic cancer patients has not yet been completely elucidated. We reported that a dipeptidyl peptidase (DPP)-4 inhibitor accelerates mammary cancer metastasis by inducing epithelial-mesenchymal transition (EMT) through the CXCL12/CXCR4/mTOR axis.
Nicolas Musi et al.
Diabetes, 51(7), 2074-2081 (2002-06-28)
Metformin is an effective hypoglycemic drug that lowers blood glucose concentrations by decreasing hepatic glucose production and increasing glucose disposal in skeletal muscle; however, the molecular site of metformin action is not well understood. AMP-activated protein kinase (AMPK) activity increases
L C Morin-Papunen et al.
The Journal of clinical endocrinology and metabolism, 85(9), 3161-3168 (2000-09-22)
Metformin, a biguanide antihyperglycemic drug, has been shown to improve ovarian function and glucose metabolism in women with polycystic ovary syndrome (PCOS), but results concerning its effects on insulin sensitivity are controversial. Oral contraceptive pills are commonly used in the
J De Jager et al.
Journal of internal medicine, 257(1), 100-109 (2004-12-21)
The UK Prospective Diabetes Study (UKPDS) showed that treatment with metformin decreases macrovascular morbidity and mortality independent of glycaemic control. We hypothesized that metformin may achieve this by improving endothelial function and chronic, low-grade inflammation. Data on this issue are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service