Skip to Content
Merck
All Photos(1)

Key Documents

IRMM447

Genomic DNA of Listeria monocytogenes (4B, NCTC 11994)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®
IRMM®

manufacturer/tradename

JRC

application(s)

genomic analysis

format

neat

storage temp.

−20°C

Analysis Note

For more information please see:
IRMM447

Legal Information

BCR is a registered trademark of European Commission
IRMM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service