Skip to Content
Merck
All Photos(1)

Key Documents

IRMM426

Wild berries (bilberries)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

IRMM®

manufacturer/tradename

JRC

format

matrix material

General description

This material is certified for the radionuclide acivity concentration of 137Cs, 90Sr and 40K.

https://ec.europa.eu/jrc/sites/default/files/rm/IRMM-426_cert.pdf
https://ec.europa.eu/jrc/sites/default/files/rm/IRMM-426_report.pdf
https://ec.europa.eu/jrc/sites/default/files/rm/IRMM-426_origin.pdf

Analysis Note

For more information please see:
IRMM426

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements: 137 Cs, 40K, 90 Sr, Aluminum (Al), Barium (Ba), Calcium (Ca), Cesium (Cs), Magnesium (Mg), Potassium (K), Rubidium (Rb), Sodium (Na), Strontium (Sr)

Matrix Group: Fruits and Vegetables

Legal Information

IRMM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service