The concentration of this product is nominally 1000 mg/L, but may vary slightly from lot to lot. The accurate concentration for each lot is on the accompanying Certificate of Analysis.
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | € 89,90 |
About This Item
grade
analytical standard
Quality Segment
CofA
current certificate can be downloaded
manufacturer/tradename
RTC ICS-034-100
technique(s)
ion chromatography: suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
single component solution
storage temp.
2-30°C
General description
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) cleaning products | application(s) cleaning products | application(s) cleaning products | application(s) cleaning products |
| format single component solution | format single component solution | format single component solution | format single component solution |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade analytical standard | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® |
| storage temp. 2-30°C | storage temp. - | storage temp. - | storage temp. - |
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the concentration of this solution?
1 answer-
Helpful?
-



