Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 mL | Check Cart for Availability | € 121,00 |
500 mL | Check Cart for Availability | € 190,00 |
About This Item
grade
certified reference material
Quality Segment
CofA
current certificate can be downloaded
manufacturer/tradename
RTC ICS-006-100
concentration
1000 μg/mL in H2O
technique(s)
ion chromatography: suitable
application(s)
environmental
food and beverages
format
single component solution
storage temp.
2-30°C
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
| format single component solution | format single component solution | format single component solution | format single component solution |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 100, 300 |
| grade certified reference material | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade analytical standard |
| storage temp. 2-30°C | storage temp. 10-30°C | storage temp. - | storage temp. 2-30°C |
Storage Class
12 - Non Combustible Liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



