Skip to Content
Merck
All Photos(1)

Key Documents

H0325000

Hexetidine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hexetidine, mixture of stereoisomers, 5-Amino-1,3-bis(2-ethylhexyl)hexahydro-5-methylpyrimidine, NSC 17764

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H45N3
CAS Number:
Molecular Weight:
339.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

hexetidine

manufacturer/tradename

EDQM

refractive index

n20/D 1.4649 (lit.)

bp

160 °C/0.4 mmHg (lit.)

density

0.889 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(CC)CN1CN(CC(CC)CCCC)CC(C)(N)C1

InChI

1S/C21H45N3/c1-6-10-12-19(8-3)14-23-16-21(5,22)17-24(18-23)15-20(9-4)13-11-7-2/h19-20H,6-18,22H2,1-5H3

InChI key

DTOUUUZOYKYHEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Involved in studies:
  • To identify pure liquid salt forms of aspirin
  • Of alkyl based selective displacers for protein purification via ion exchange chromatography
  • Of the in vivo application of models for testing drugs against nonreplicating Mycobacterium tuberculosis
  • Nutrient-sensitized screening for drugs that shift the energy metabolism from mitochondrial respiration to glycolysis
  • Inhibition of angiogensis
  • Screening antimalarial drugs

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Peyrl et al.
Journal of neuro-oncology, 120(1), 139-145 (2014-07-16)
The Ommaya reservoir facilitates repetitive delivery of drugs into the CSF and is a pharmacologically rational system for intrathecal chemotherapy. Because previous studies have found a high rate of infection and other complications we herein studied our experience with this
Elvedina Kapić et al.
Medicinski arhiv, 56(1), 43-48 (2002-03-29)
Hexetidine is very safe oral antiseptic with broad antibacterial and antifungal activity in vivo and in vitro. It has local-anesthetics, astringent and deodorant activity. Also, it has very strong antiplac effects. Resistention of microorganisms on hexetidine is short and transient.
F Afennich et al.
International journal of dental hygiene, 9(3), 182-190 (2011-03-02)
To review the literature concerning hexetidine-containing mouthwash as a monotherapy or as an adjunct to oral hygiene in the prevention of plaque accumulation and gingival inflammation. PubMed-MEDLINE and the Cochrane-CENTRAL were searched through January 2010 to identify appropriate studies. The
Synergistic antiplaque effects of a zinc fluoride/hexetidine containing mouthwash. A review.
U P Saxer et al.
Schweizerische Monatsschrift fur Zahnheilkunde = Revue mensuelle suisse d'odonto-stomatologie, 93(8), 689-704 (1983-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service