Skip to Content
Merck
All Photos(1)

Key Documents

GSK5305

Supelco

4-AMINOPHENOL

Sign Into View Organizational & Contract Pricing

Select a Size

GSK5305-1G
€7.60

€7.60


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
GSK5305-1G
€7.60

About This Item

UNSPSC Code:
41116107

€7.60


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service