Skip to Content
Merck

Skip To

FPCOC1

Flash Point Standard - Cleveland Open Cup

certified reference material, ASTM D 92 (85 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 x 80 mL
Check Cart for Availability
€ 252,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904

€ 252,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

according to ASTM® D92

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum
pharmaceutical

format

neat

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Cleveland Open Cup Flash Point Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Application

Flash point standard-cleveland open cup may be used as a standard in determining the flash point of mineral base oils, which may be used as a parameter for testing the biodegradability of mineral base oils, using CEC L-33-A-93 test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
FPCOC3FPCOC4FPCOC5
description

certified reference material, ASTM D 92 (85 °C)

description

certified reference material, ASTM D 92 (148 °C)

description

certified reference material, ASTM D 92 (188 °C)

description

certified reference material, ASTM D 92 (257 °C)

format

neat

format

neat

format

neat

format

neat

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

application(s)

petroleum
pharmaceutical

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label


Still not finding the right product?


Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

10 - Combustible liquids

Flash Point (°F)

168.8 °F

Flash Point (°C)

76 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters