Skip to Content
Merck
All Photos(1)

Key Documents

F8639

Supelco

Famprofazone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31N3O
CAS Number:
Molecular Weight:
377.52
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CC(C)C1=C(CN(C)C(C)Cc2ccccc2)N(C)N(c3ccccc3)C1=O

InChI

1S/C24H31N3O/c1-18(2)23-22(17-25(4)19(3)16-20-12-8-6-9-13-20)26(5)27(24(23)28)21-14-10-7-11-15-21/h6-15,18-19H,16-17H2,1-5H3

InChI key

GNUXVOXXWGNPIV-UHFFFAOYSA-N

General description

Famprofazone is a pyrazole derivative and belongs to the class of non-steroidal anti-inflammatory drugs. It is widely used as an analgesic and antipyretic in medical applications.

Application

Famprofazone may be used as an analytical reference standard for the quantification of the analyte in biological samples using liquid chromatography coupled to tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brandy Greenhill et al.
Journal of analytical toxicology, 27(7), 479-484 (2003-11-11)
There are a several drugs that lead to the production of methamphetamine and/or amphetamine in the body which are subsequently excreted in the urine. These drugs raise obvious concerns when interpreting positive amphetamine drug testing results. Famprofazone is an analgesic
F Musshoff et al.
International journal of legal medicine, 111(6), 305-308 (1998-11-24)
After a traffic accident a 32-year-old man was suspected of having previously taken an illegal drug. An immunochemical screening procedure revealed positive results for amphetamines in both urine and blood samples. The preliminary test was confirmed by GC/MS and both
H S Shin
Chirality, 9(1), 52-58 (1997-01-01)
Following administration of famprofazone to humans, the stereoselective metabolism from the drug to its known metabolites (+,-)-ephedrine, (+,-)-pseudoephedrine, (+,-)-norephedrine, (+,-)-norpseudoephedrine, (+,-)-p-hyroxyamphetamine, (+,-)-p-hydroxymethamphetamine, and (+,-)-p-hydroxynorephedrine was studied. The enantiomers of the metabolites were derivatized with alpha-methoxy-alpha -(trifluoromethyl)-phenylacetyl chloride (MPTA.Cl) as the
A rapid screening LC?MS/MS method based on conventional HPLC pumps for the analysis of low molecular weight xenobiotics: application to doping control analysis
Mazzarino M, et al.
Drug Testing and Analysis, 2(7), 311-322 (2010)
Forensic Science
Handbook of Analytical Chemistry, 6(7), 311-322 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service