Skip to Content
Merck
All Photos(1)

Key Documents

F0763

Sigma-Aldrich

Fura 2 pentapotassium salt

≥80% purity, powder

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€474.00

€474.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€474.00

About This Item

Empirical Formula (Hill Notation):
C29H22K5N3O14
CAS Number:
Molecular Weight:
831.99
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€474.00


Estimated to ship on18 May 2025


Request a Bulk Order

Product Name

Fura 2 pentapotassium salt, ≥80%

Quality Level

Assay

≥80%

form

powder

color

yellow

solubility

1 M NaOH: 10 mg/mL

εmax

24-37 at 363 nm (pH 7.2)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[K+].[K+].[K+].[K+].[K+].Cc1ccc(N(CC([O-])=O)CC([O-])=O)c(OCCOc2cc3cc(oc3cc2N(CC([O-])=O)CC([O-])=O)-c4ncc(o4)C([O-])=O)c1

InChI

1S/C29H27N3O14.5K/c1-15-2-3-17(31(11-24(33)34)12-25(35)36)20(6-15)43-4-5-44-21-7-16-8-22(28-30-10-23(46-28)29(41)42)45-19(16)9-18(21)32(13-26(37)38)14-27(39)40;;;;;/h2-3,6-10H,4-5,11-14H2,1H3,(H,33,34)(H,35,36)(H,37,38)(H,39,40)(H,41,42);;;;;/q;5*+1/p-5

InChI key

HQKWBBCTCLUGSX-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

General description

Fura 2 pentapotassium salt (fura-2) is a fluorescent ratiometric Ca2+ dye.[1][2] Fura 2 pentapotassium salt (fura-2) helps to estimate the level of cytosolic Ca(2+) by determining the emission found at 510 nm after alternating excitation at 340 nm and 380 nm (F340/F380 ratio).[1] Hence, it is considered as the standard for quantitative intracellular calcium measurements.[2]

Application

Cell impermeable fluorescent probe for Ca2+.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resveratrol interferes with fura-2 intracellular calcium measurements
Kopp, et al.
Journal of Fluorescence, 24(2), 279-284 (2014)
Ram Chandra Paudel et al.
Biochemical and biophysical research communications, 450(4), 1626-1630 (2014-07-19)
Fura-2 is a commonly used fluorescent Ca(2+) dye that allows an accurate determination of cytosolic Ca(2+) levels by measuring the emission obtained at 510 nm after alternating excitation at 340 nm and 380 nm (F340/F380 ratio). Previous studies, based on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service