Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Check Cart for Availability | € 145,00 |
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
grade
certified reference material
agency
ERM®
manufacturer/tradename
JRC
application(s)
petroleum
format
matrix material
Analysis Note
A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage
please visit the JRC webpage
Legal Information
ERM is a registered trademark of European Commission
1 of 1
This Item | |||
|---|---|---|---|
| description ERM®, certified reference material | description ERM®, certified reference material | description Gas Oil (Butoxybenzene) | description ERM®, certified reference material |
| grade certified reference material | grade certified reference material | grade - | grade certified reference material |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| application(s) petroleum | application(s) petroleum | application(s) petroleum | application(s) petroleum |
| format matrix material | format matrix material | format - | format matrix material |
| agency ERM® | agency ERM® | agency - | agency ERM® |
Still not finding the right product?
Explore all of our products under Diesel (B7)
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2
Storage Class
3 - Flammable liquids
WGK
WGK 3
Flash Point (°F)
134.6 °F
Flash Point (°C)
57 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




