Skip to Content
Merck
All Photos(1)

Key Documents

ERMDA476IFCC

Human serum (anti-MPO IgG)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Agency

ERM®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

storage temp.

−70°C

General description

Certified for the mass concentration of anti-myeloperoxidase immunoglobulin G in human serum.

ERM-DA476IFCC_cert

ERM-DA476IFCC_report

ERM-DA476IFCC_origin

Analysis Note

For more information please see:
ERMDA476IFCC

Legal Information

ERM is a registered trademark of European Commission

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service