Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
14 g | Check Cart for Availability | € 323,00 |
About This Item
grade
certified reference material
agency
ERM®, certified by the BSC
description
Storage Temp--35 to -45 °C
composition
dibutyltin (DBT), 1.54 mg/kg ±0.12 mg/kg by , monobutyltin (MBT), 1.50 mg/kg ±0.28 mg/kg by , tributyltin (TBT), 2.2 mg/kg ±0.19 mg/kg by
manufacturer/tradename
JRC
matrix
mussel
application(s)
food and beverages
format
matrix material
shipped in
wet ice
Packaging
Analysis Note
ERMCE477
Other Notes
Trace Elements / Metallorganics: DBT: Sn(C4H9)2, MBT: Sn(C4H9)3, TBT: Sn(C4H9)3
Matrix Group: Fish and Seafood
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) food and beverages | application(s) - | application(s) food and beverages | application(s) food and beverages |
| format matrix material | format matrix material | format matrix material | format matrix material |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. - |
| manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC | manufacturer/tradename JRC |
| composition dibutyltin (DBT), 1.54 mg/kg ±0.12 mg/kg by , tributyltin (TBT), 2.2 mg/kg ±0.19 mg/kg by , monobutyltin (MBT), 1.50 mg/kg ±0.28 mg/kg by | composition - | composition - | composition - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
