Skip to Content
Merck

Skip To

ERMCA615

Ground water (trace elements)

ERM®, certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mL
Check Cart for Availability
€ 221,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 221,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Analysis Note

A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:
please visit the JRC webpage

Legal Information

ERM is a registered trademark of European Commission

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BCR320RBCR701BCR142R
description

ERM®, certified reference material

description

BCR®, certified reference material

description

BCR®, certified reference material

description

BCR®, certified reference material

manufacturer/tradename

JRC

manufacturer/tradename

JRC

manufacturer/tradename

JRC

manufacturer/tradename

JRC

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

matrix material

format

matrix material

format

matrix material

format

matrix material

application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

agency

ERM®

agency

BCR®

agency

BCR®

agency

BCR®


Storage Class

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

10325-94-7 + 10099-74-8 + 10045-94-0

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters