Skip to Content
Merck

Skip To

ERMCA100

Water (PAHs)

ERM®, certified reference material

Synonym(s):

Surface Water

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 423,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 423,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

ERM®

manufacturer/tradename

JRC

application(s)

environmental

format

matrix material

Analysis Note

A JRC Certificate of Analysis is included with the product, a pdf copy of the CoA and other documentation you can find here:

please visit the JRC webpage

Legal Information

ERM is a registered trademark of European Commission

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ERMCA616ERMCZ130ERMEF317
description

ERM®, certified reference material

description

ERM®, certified reference material

description

ERM®, certified reference material

description

ERM®, certified reference material

application(s)

environmental

application(s)

environmental

application(s)

environmental
industrial qc

application(s)

petroleum

format

matrix material

format

matrix material

format

matrix material

format

matrix material

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

manufacturer/tradename

JRC

manufacturer/tradename

JRC

manufacturer/tradename

JRC

manufacturer/tradename

JRC

agency

ERM®

agency

ERM®

agency

ERM®

agency

ERM®


Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters