Skip to Content
Merck
All Photos(1)

Key Documents

ERMBB184

Bovine muscle (trace elements)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

7 G
€289.00

€289.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
7 G
€289.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

€289.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

format

matrix material

General description

Certified for the mass fractions of As, Cd, Cu, Fe, Mn, Se, Zn

Analysis Note

For more information please see:
ERMBB184

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements: Arsenic (As), Cadmium (Cd), Calcium (Ca), Chloride, Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Potassium (K), Selenium (Se), Sodium (Na), Zinc (Zn)

Matrix Group: Meat

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service