Skip to Content
Merck
All Photos(1)

Key Documents

ERMAC699

Zearalenone in acetonitrile

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

4 ML
€185.00

€185.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
4 ML
€185.00

About This Item

EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

€185.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

manufacturer/tradename

JRC

application(s)

agriculture
general analytical

format

single component solution

storage temp.

2-8°C

Analysis Note

For more information please see:
ERMAC699

Legal Information

ERM is a registered trademark of European Commission

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service