Skip to Content
Merck
All Photos(1)

Key Documents

DLW354451

Corning® BioCoat Dish 100mm Fibronectin 5pac 10cas

Synonym(s):

plastic cell culture dish

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812

shipped in

wet ice

storage temp.

2-8°C

Legal Information

BioCoat is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service