Skip to Content
Merck
All Photos(1)

Documents

D2954000

Dobutamine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-3,4-Dihydroxy-N-[3-(4-hydroxyphenyl)-1-methylpropyl]-β-phenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO3 · HCl
CAS Number:
Molecular Weight:
337.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dobutamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CC(CCc1ccc(O)cc1)NCCc2ccc(O)c(O)c2

InChI

1S/C18H23NO3.ClH/c1-13(2-3-14-4-7-16(20)8-5-14)19-11-10-15-6-9-17(21)18(22)12-15;/h4-9,12-13,19-22H,2-3,10-11H2,1H3;1H

InChI key

BQKADKWNRWCIJL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dobutamine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin G Schultz et al.
American journal of hypertension, 26(4), 557-566 (2013-03-08)
The J-curve relationship between brachial diastolic blood pressure (DBP) and mortality is believed to be mediated through reduced myocardial perfusion. This study aimed to determine the relationship between DBP and subendocardial perfusion in patients with and without coronary artery disease
Neeraj Singh et al.
Transplantation, 94(12), 1224-1229 (2012-12-25)
The utility of cardiac stress testing as a risk-stratification tool before kidney transplantation remains debatable owing to discordance with coronary angiography and outcome yields at different centers. We conducted a retrospective study of 273 diabetic kidney transplant recipients from 2006
X Monnet et al.
British journal of anaesthesia, 111(5), 743-749 (2013-06-27)
We tested the ability of mean acceleration (Acc) and peak velocity (V peak) of the aortic velocity signal measured by oesophageal Doppler to reflect left ventricular (LV) systolic performance. We included critically ill patients in whom a fluid challenge (n=25)
Kiril A Ahtarovski et al.
American journal of physiology. Heart and circulatory physiology, 305(7), H1098-H1103 (2013-07-03)
Men and women are known to react differently to stress. Thus, stress cardiomyopathy almost solely strikes women. Stress cardiomyopathy is suggested to relate to sex differences in catecholamine reaction. Left heart function during dobutamine stress is well described, but sex-specific
Marie-Annick Clavel et al.
JACC. Cardiovascular imaging, 6(2), 175-183 (2013-03-16)
The objective of this study was to examine the value of stress-echocardiography in patients with paradoxical low-flow, low-gradient (PLFLG) aortic stenosis (AS). The projected aortic valve area (AVAProj) at a normal flow rate was calculated in 55 patients with PLFLG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service