Skip to Content
Merck
All Photos(1)

Key Documents

CRMADKR

Acidity Standard

ASTM D 3242 (0.0021 mg KOH/g)

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€304.00

€304.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€304.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€304.00


Estimated to ship on29 May 2025


Request a Bulk Order

grade

certified reference material

Agency

according to ASTM® D3242

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
petroleum

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Fuels Testing Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM)

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 - 158.0 °F - closed cup

Flash Point(C)

29 - 70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service