Skip to Content
Merck
All Photos(1)

Key Documents

CRM016

Supelco

Trace Metals - Fresh Water Sediment 3

certified reference material, pkg of 50 g

Synonym(s):

Sediment matrix CRM, trace metals

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€218.00

€218.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
50 G
€218.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

€218.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM016-050

technique(s)

AAS: suitable
ICP: suitable

pH

7.85

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application


  • Bioremediation of AMD-polluted areas: The study examines the response of Propsilocerus akamusi to leachates from AMD-contaminated sediments using trace metal analysis, emphasizing the potential of metal bioremediation in aquatic environments, which is crucial for maintaining ecosystem health and supporting biotechnological applications in pharma and biotech industries (Zheng et al., 2024).

  • Marine pollution control using dead seaweeds: Research on metal uptake by dead seaweeds provides insights into controlling marine pollution, highlighting the role of trace metal analysis in assessing environmental impacts and improving strategies for the pharma and biotech sectors (Vázquez-Arias et al., 2023).

  • Mercury source tracing in coastal lakes: This study traces mercury sources and depositional history in northeastern U.S. lakes, utilizing trace metal analysis techniques, which are pivotal in environmental monitoring and have implications for public health and pharmaceutical research (Taylor et al., 2022).

  • Metal bioaccumulation in urban river habitats: An assessment of metal bioaccumulation in freshwater gastropods from urban river habitats provides valuable data for environmental and pharmaceutical research, emphasizing the importance of trace metal analysis in studying biogeochemical cycling (Chukaeva and Petrov, 2023).

  • Health hazards of elements in freshwater crayfish: The bioaccumulation and biosedimentation study of elements in crayfish from the River Nile addresses health hazards related to heavy metal contamination, serving as a critical reference for environmental safety and pharmacological studies (Abbas et al., 2023).

Other Notes

The following elements are not certified and are included for information only:
  • Sb, B, Mo, Se, Si, Ag, Sr, Tl
Values of analytes vary lot to lot. Some lots may not contain all analytes.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Analyte

Description

    Aluminum
    Antimony
    Arsenic
    Barium
    Beryllium
    Boron
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silicon
    Silver
    Sodium
    Strontium
    Thallium
    Vanadium
    Zinc
    See All (27)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

EU REACH Annex XIV (Authorisation List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service