BP296
Procyclidine hydrochloride
British Pharmacopoeia (BP) Reference Standard
Synonym(s):
α-Cyclohexyl-α-phenyl-1-pyrrolidinepropanol
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
procyclidine
form
solid
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
BP
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
storage temp.
2-8°C
SMILES string
Cl.OC(CCN1CCCC1)(C2CCCCC2)c3ccccc3
InChI
1S/C19H29NO.ClH/c21-19(17-9-3-1-4-10-17,18-11-5-2-6-12-18)13-16-20-14-7-8-15-20;/h1,3-4,9-10,18,21H,2,5-8,11-16H2;1H
InChI key
ZFSPFXJSEHCTTR-UHFFFAOYSA-N
Gene Information
human ... CHRM1(1128)
Looking for similar products? Visit Product Comparison Guide
General description
Application
Also used in monographs such as:
Packaging
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service