Skip to Content
Merck
All Photos(1)

Documents

BCR352

ZnAl4 (trace elements)

BCR®, certified reference material, disc

Synonym(s):

Aluminum-zinc alloy, Zinc-aluminum alloy Zn with 4% Al

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

form

disc

manufacturer/tradename

JRC

application(s)

general analytical

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR352

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service