Skip to Content
Merck
All Photos(1)

Key Documents

BCR307

2-Nitronaphthalene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO2
CAS Number:
Molecular Weight:
173.17
Beilstein:
2046354
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

65-73 °C

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2ccccc2c1

InChI

1S/C10H7NO2/c12-11(13)10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

ZJYJZEAJZXVAMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR307

Legal Information

BCR is a registered trademark of European Commission

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

all-trans-retinol modulation of nitronaphthalene-induced lung and liver injury in male Sprague-Dawley rats.
J M Sauer et al.
Proceedings of the Western Pharmacology Society, 38, 29-31 (1995-01-01)
Karolina Pecková et al.
Analytical and bioanalytical chemistry, 381(2), 520-525 (2004-12-31)
The polarographic behaviour of 2-nitronaphthalene was investigated by DC tast polarography (DCTP) and differential pulse polarography (DPP), both at a dropping mercury electrode, and differential pulse voltammetry and adsorptive stripping voltammetry, both at a hanging mercury drop electrode. Optimum conditions
M Watanabe et al.
Mutation research, 301(1), 7-12 (1993-01-01)
Salmonella typhimurium YG1024 is a derivative of S. typhimurium TA98 with a high level of N-hydroxyarylamine O-acetyltransferase (OAT) activity. We have demonstrated that this strain is highly sensitive to the mutagenic actions of N-hydroxyarylamines derived from aromatic amines and nitroarenes.
J M Sauer et al.
Toxicology, 105(2-3), 237-249 (1995-12-28)
We have previously shown that retinol pretreatment limits the amount of pulmonary injury caused by 1-nitronaphthalene in male Sprague-Dawley rats. The main objective of this study was to determine if retinol pretreatment can protect the lung from the toxicity of
J C Sasaki et al.
Mutation research, 445(1), 113-125 (1999-10-16)
2-Nitronaphthalene (2NN) has been identified as a mutagenic atmospheric reaction product of naphthalene in the Ames bacterial reversion assay. Recent experiments have shown this nitroarene to be genotoxic in a human lymphoblastoid cell line (MCL-5) transfected with plasmids encoding epoxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service