Skip to Content
Merck
All Photos(1)

Documents

BCR157

Benz[a]acridine

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H11N
CAS Number:
Molecular Weight:
229.28
Beilstein:
9262
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

c1ccc2nc3ccc4ccccc4c3cc2c1

InChI

1S/C17H11N/c1-3-7-14-12(5-1)9-10-17-15(14)11-13-6-2-4-8-16(13)18-17/h1-11H

InChI key

JEGZRTMZYUDVBF-UHFFFAOYSA-N

Analysis Note

For more information please see:
BCR157

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Motohashi et al.
Anticancer research, 12(4), 1207-1210 (1992-07-01)
The abilities of 14 phenothiazines, 8 benzo[a]phenothiazines and 12 benz[c]acridines to induce an antibacterial effect against Escherichia coli K12 were compared. Several phenothiazines, which showed antiplasmid activity, displayed the most potent antibacterial activity. All benz[c]acridine derivatives were moderately antibacterial, whereas
Benz[a]acridine.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 32, 123-127 (1983-12-01)
J Jacob et al.
Cancer letters, 16(3), 297-306 (1982-09-01)
The metabolism of benz[a]- and benz[c]acridine by liver and lung microsomes from untreated, phenobarbital (PB)-treated and benzo[k]fluoranthene (BkF)-treated rats has been studied by gas chromatography/mass spectrometry (GC/MS). Epoxidation and hydrolysis of the epoxides to dihydrodiols were found to be the
R P Deutsch-Wenzel et al.
Cancer letters, 20(1), 97-101 (1983-08-01)
Using a beeswax/tricaprylin mixture as vehicle, three doses each of acridine, benz[a]acridine (BaAC), benz[c]acridine (BcAC), dibenz[a,h]-acridine (DBa,hAC) and dibenz[a,j]acridine (DBa,jAC) were injected into the lungs of 35 female Osborne-Mendel rats per group. To compare the carcinogenic potency of the heterocycles
J Molnár et al.
Anticancer research, 13(4), 1019-1025 (1993-07-01)
This review summarizes our experiments which are investigating the relationship between the structure and activity of mainly phenothiazines, benzo[a]phenothiazines and benz[c]acridines. Phenothiazines had potent antiplasmid and antibacterial activities, but induced weak antimicrobial activity in vivo. Their antiplasmid activity seemed to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service