Skip to Content
Merck
All Photos(2)

Key Documents

A42409

Sigma-Aldrich

2-Aminobiphenyl

≥96.5% purity, powder or crystals

Synonym(s):

2-Biphenylylamine, 2-Phenylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€95.90

€95.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€95.90

About This Item

Linear Formula:
C6H5C6H4NH2
CAS Number:
Molecular Weight:
169.22
Beilstein:
471874
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€95.90


Check Cart for Availability

Request a Bulk Order

Product Name

2-Aminobiphenyl, ≥96.5% (GC)

vapor density

5.8 (vs air)

Quality Level

Assay

≥96.5% (GC)

form

powder or crystals

autoignition temp.

842 °F

technique(s)

titration: suitable

impurities

<0.1% 4-aminobiphenyl

color

brown
off-white to yellow

bp

299 °C (lit.)

mp

47-50 °C (lit.)

solubility

water: insoluble

density

1.16 g/cm3 at 21 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Nc1ccccc1-c2ccccc2

InChI

1S/C12H11N/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9H,13H2

InChI key

TWBPWBPGNQWFSJ-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

General description

2-Aminobiphenyl is a substrate for UGTs (UDP-glucuronosyltransferases), including UGT1A4, UGT2B13 and UGT2B16.[1]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

307.4 °F - closed cup

Flash Point(C)

153 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Cheng Chen et al.
Chemical research in toxicology, 25(3), 695-705 (2012-02-01)
Overexposure to biphenyl amine compounds, which are found in smoke and azo-dyes, is linked to the occurrence of bladder cancer. However, the molecular mechanisms of biphenyl amine compound-induced bladder cancer are still unclear. Many studies have demonstrated that overexpression of
Shu Chi Wang et al.
Mutation research, 593(1-2), 9-21 (2005-08-23)
4-Aminobiphenyl (4-ABP) and its analogue, 2-aminobiphenyl (2-ABP), were examined for their ability to induce oxidative DNA damage in Hep G2 cells. Using the alkaline comet assay, we showed that 2-ABP and 4-ABP (25-200 microM) were able to induce the DNA
High-performance liquid chromatography with on-line radioactivity monitoring for metabolic studies of 2- and 4-[14C]aminobiphenyls.
S D Jatoe et al.
Journal of chromatography, 442, 394-400 (1988-06-17)
K M Abdo et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 2(5), 201-210 (1982-09-01)
Diets containing 0.1 or 0.3% 2-biphenylamine hydrochloride were fed to groups of 50 Fischer 344 rats and 50 B6C3F1 mice of each sex for 104-106 weeks. Mean body weights of high-dose rats of both sexes and of low-dose male rats
Agricultural and Biological Chemistry, 38, 1209-1209 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service