Skip to Content
Merck
All Photos(1)

Key Documents

A2161

Supelco

Amino acid standards for fluorescence detection

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€108.00

€108.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
10 ML
€108.00

About This Item

UNSPSC Code:
85151701
eCl@ss:
32160406
NACRES:
NA.24

€108.00


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

suitable for fluorescence detection
Qualitative standard. Amino acids and related compounds are in 0.1N HCl.

Analyte

Description

    L-Alanine
    Ammonium chloride
    L-Arginine
    L-Aspartic acid
    L-Cystine
    L-Glutamic acid
    Glycine
    L-Histidine
    L-Isoleucine
    L-Leucine
    L-Lysine
    L-Methionine
    L-Phenylalanine
    L-Proline
    L-Serine
    L-Threonine
    L-Tyrosine
    L-Valine
    See All (18)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Somaieh Zafari et al.
Plant physiology and biochemistry : PPB, 99, 11-20 (2015-12-29)
Lead (Pb) is a hazardous heavy metal present in the environment which elicits oxidative stress in plants. To characterize the physiological and biochemical basis of Pb tolerance, Prosopis farcta seedlings were exposed to Hoagland's solutions at six different Pb concentrations
Hannaneh Tashackori et al.
Journal of plant physiology, 232, 115-126 (2018-12-12)
Lignans are diphenolic compounds produced in plants via coupling of two coniferyl alcohol molecules with the aid of a dirigent protein to form pinoresinol (PINO). The latter is reduced via lariciresinol (LARI) to secoisolariciresinol by the bifunctional pinoresinol-lariciresinol reductase (PLR).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service