Skip to Content
Merck

Skip To

A2134

4-Aminomethylbenzenesulfonamide hydrochloride

analytical standard, ≥95%

Synonym(s):

4-Homosulfanilamide, Mafenide, Marfanil

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Check Cart for Availability
€ 255,00

About This Item

Empirical Formula (Hill Notation):
C7H10N2O2S · HCl
CAS Number:
Molecular Weight:
222.69
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
205-325-3
MDL number:

€ 255,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95%

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.NCc1ccc(cc1)S(N)(=O)=O

InChI

1S/C7H10N2O2S.ClH/c8-5-6-1-3-7(4-2-6)12(9,10)11;/h1-4H,5,8H2,(H2,9,10,11);1H

InChI key

SIACJRVYIPXFKS-UHFFFAOYSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2799T17633984
grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

format

neat

format

neat

format

neat

format

neat

Gene Information

human ... CA1(759), CA2(760), CA5A(763), CA5B(11238), CA9(768)
mouse ... Car13(71934), Car5a(12352)

Gene Information

-

Gene Information

human ... ESR1(2099), ESR2(2100), ESRRG(2104), IL6(3569)
rat ... Ar(24208), Esr1(24890), Esr2(25149)

Gene Information

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

assay

≥95%

assay

-

assay

-

assay

-

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters