Skip to Content
Merck
All Photos(1)

Key Documents

99314

Supelco

Sodium ionophore I - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

2-Nitrophenyl octyl ether, 89.5 wt. % (73732)
Sodium ionophore I, 10.0 wt. % (71732)
Sodium tetraphenylborate, 0.5 wt. % (72018)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lydie Cheval et al.
American journal of physiology. Renal physiology, 317(2), F435-F443 (2019-06-13)
We have recently reported that type A intercalated cells of the collecting duct secrete Na+ by a mechanism coupling the basolateral type 1 Na+-K+-2Cl- cotransporter with apical type 2 H+-K+-ATPase (HKA2) functioning under its Na+/K+ exchange mode. The first aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service